Novus Biologicals
Manufacturer Code:NBP234720AF750
Catalog # NP234720A70
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | TA99 |
Immunogen | SK-MEL-23 cells |
Conjugate | Alexa Fluor 750 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: 5,6-dihydroxyindole-2-carboxylic acid oxidase; b-PROTEIN CAS256-dihydroxyindole-2-carboxylic acid oxidase Catalase B CATB EC 1.14.18 EC 1.14.18.- EC 1.14.18.1 Glycoprotein 75 GP75DHICA oxidase Melanoma antigen gp75 TRP-1 TRPTYRP tyrosinase-related protein 1TRP1 TYRRP; Catalase B; DHICA oxidase; Glycoprotein 75; Melanoma antigen gp75; TRP; Tyrosinase-related protein 1
Gene Aliases: b-PROTEIN; CAS2; CATB; GP75; OCA3; TRP; TRP1; TYRP; TYRP1; TYRRP
UniProt ID: (Human) P17643
Entrez Gene ID: (Human) 7306
Molecular Function:
oxidase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.