Invitrogen
Manufacturer Code:MA534621
Catalog # PIMA534621
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JU36-48 |
Immunogen | Recombinant protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles store in dark |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: 5,6-dihydroxyindole-2-carboxylic acid oxidase; 56-dihydroxyindole-2-carboxylic acid oxidase B BOS_8760 b-PROTEIN BROWN Brown locus protein CAS2 Catalase B CATB DHICA oxidase Glycoprotein 75 GP75 isa Melanoma antigen gp75 MGC120175 MGC149362 OCA3 RP11-3L8.1 TRP TRP1 TRP-1 TRY1 TRY4 TRYP1 Tyrosinase-related protein 1 TYRP Tyrp1 Tyrp-1 TYRRP; Catalase B; DHICA oxidase; Glycoprotein 75; Melanoma antigen gp75; TRP; Tyrosinase-related protein 1
Gene Aliases: b-PROTEIN; CAS2; CATB; GP75; OCA3; TRP; TRP1; TYRP; TYRP1; TYRRP
UniProt ID: (Human) P17643
Entrez Gene ID: (Human) 7306
Molecular Function: oxidase oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.