Invitrogen
Manufacturer Code:PA569298
Catalog # PIPA569298
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of rat TYRP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5,6-dihydroxyindole-2-carboxylic acid oxidase; 56-dihydroxyindole-2-carboxylic acid oxidase DHICA oxidaseTRP-1 TRP1 catalase B glycoprotein 75 melanoma antigen gp75; Brown locus protein; Catalase B; DHICA oxidase; Glycoprotein 75; gp75; iris stromal atrophy; melanogenic enzyme; Melanoma antigen gp75; TRP; TRP-1; TRP1; truncated Tyrp1; Tyrosinase-related protein 1; tyrosine phosphatase 1, same as B (Brown); TYRP1
Gene Aliases: B; b-PROTEIN; BROWN; CAS2; CATB; GP75; isa; OCA3; TRP; TRP-1; TRP1; TYRP; Tyrp-1; TYRP1; TYRRP
UniProt ID: (Bovine) Q8WN57, (Human) P17643, (Mouse) P07147
Entrez Gene ID: (Rabbit) 100342170, (Bovine) 282105, (Sheep) 100034666, (Dog) 403479, (Horse) 100033886, (Goat) 100861390, (Human) 7306, (Guinea pig) 100717475, (Rat) 298182, (Mouse) 22178
Molecular Function: oxidase oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.