Invitrogen
Manufacturer Code:PA5107304
Catalog # PIPA5107304
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide located within the following region: NDPIFVLLHT FTDAVFDEWL RRYNADISTF PLENAPIGHN RQYNMVPFWP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5,6-dihydroxyindole-2-carboxylic acid oxidase; Brown locus protein; Catalase B; DHICA oxidase; Glycoprotein 75; iris stromal atrophy; Melanoma antigen gp75; TRP; Tyrosinase-related protein 1
Gene Aliases: b; b-PROTEIN; brown; CAS2; CATB; GP75; isa; OCA3; TRP; TRP-1; TRP1; TYRP; Tyrp-1; TYRP1; TYRRP
UniProt ID: (Human) P17643, (Mouse) P07147
Entrez Gene ID: (Human) 7306, (Mouse) 22178
Molecular Function:
oxidase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.