Novus Biologicals
Manufacturer Code:NBP180653
Catalog # NBP180653
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:ECTSFIPEARAVLDLVDQCPKQIQKGKFQVVAIEGLDATGKTTVTQSVADSLKAVLLKSPPSCIGQWRKIFDDEPTIIRRAFYSLGNYIVASEIAKESAKSPV |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cytidine monophosphate (UMP-CMP) kinase 2 mitochondrial cytidylate kinase 2 EC 2.7.4.14 mitochondrial mitochondrial UMP-CMP kinase thymidine monophosphate kinase 2 thymidylate kinase family LPS-inducible member TMPK2 TYKi UMP-CMPK2; cytidine monophosphate (UMP-CMP) kinase 2, mitochondrial; cytidylate kinase 2; mitochondrial UMP-CMP kinase; Nucleoside-diphosphate kinase; thymidine monophosphate kinase 2; thymidylate kinase family LPS-inducible member; UMP-CMP kinase 2, mitochondrial; UMP/CMP kinase
Gene Aliases: CMPK2; NDK; TMPK2; TYKi; UMP-CMPK2
UniProt ID: (Human) Q5EBM0
Entrez Gene ID: (Human) 129607
Molecular Function:
kinase
nucleotide kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.