OriGene
Manufacturer Code:TA811366
Catalog # 50-168-3996
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI7B2 |
Immunogen | Full length human recombit protein of human TXNRD1 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: gene associated with retinoic and IFN-induced mortality 12 protein; Gene associated with retinoic and interferon-induced mortality 12 protein; GRIM-12; GRIM-12 TR TR1 TRXR1 TXNR; KM-102-derived reductase-like factor; NADPH-dependent thioredoxin reductase; oxidoreductase; selenoprotein oxidoreductase; testis tissue sperm-binding protein Li 46a; Thioredoxin reductase 1, cytoplasmic; thioredoxin reductase GRIM-12; Thioredoxin reductase TR1; TR; TR alpha
Gene Aliases: GRIM-12; GRIM12; KDRF; TR; TR1; TRXR1; TXNR; TXNRD1
UniProt ID: (Human) Q6VBP2, (Mouse) Q9JMH6
Entrez Gene ID: (Human) 7296, (Rat) 58819, (Mouse) 50493
Molecular Function: dehydrogenase oxidase oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.