Invitrogen
Manufacturer Code:PA576132
Catalog # PIPA576132
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein of human TXNL4A. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DIM1 protein homolog; Spliceosomal U5 snRNP-specific 15 kDa protein; thioredoxin-like 4; thioredoxin-like 4A; Thioredoxin-like protein 4A; Thioredoxin-like U5 snRNP protein U5-15kD; TXNL4A DIM1 TXNL4 Thioredoxin-like protein 4A
Gene Aliases: BMKS; D18Wsu98e; DIB1; DIM1; ENSMUSG00000057130; SNRNP15; TXNL4; TXNL4A; U5-15kD; U5-15kDa
UniProt ID: (Human) P83876, (Mouse) P83877
Entrez Gene ID: (Human) 10907, (Mouse) 27366
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.