Novus Biologicals
Manufacturer Code:NBP299990AF350
Catalog # NB090555
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant TXNL4A (P83876 Met1-Tyr142). |
Conjugate | Alexa Fluor 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DIB1 DIM1 protein homolog DIM1Thioredoxin-like U5 snRNP protein U5-15kD HsT161 Spliceosomal U5 snRNP-specific 15 kDa protein thioredoxin-like 4 thioredoxin-like 4A thioredoxin-like protein 4A TXNL4 U5-15kD; DIM1 protein homolog; Spliceosomal U5 snRNP-specific 15 kDa protein; thioredoxin-like 4; thioredoxin-like 4A; Thioredoxin-like protein 4A; Thioredoxin-like U5 snRNP protein U5-15kD
Gene Aliases: BMKS; DIB1; DIM1; SNRNP15; TXNL4; TXNL4A; U5-15kD
UniProt ID: (Human) P83876
Entrez Gene ID: (Human) 10907
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.