Invitrogen
Manufacturer Code:PA5115291
Catalog # PIPA5115291
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human GLRX3 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bA500G10.4 FLJ11864 GLRX3 GLRX4 glutaredoxin 3 glutaredoxin 4 glutaredoxin-3 GRX3 GRX4 HUSSY-22 PICOT PKC interacting cousin of thioredoxin PKC-interacting cousin of thioredoxin PKCq-interacting protein PKC-theta-interacting protein testicular tissue protein Li 75 thioredoxin-like 2 thioredoxin-like protein 2 TXNL2 TXNL3; glutaredoxin 4; Glutaredoxin-3; PICOT; PKC interacting cousin of thioredoxin; PKC-interacting cousin of thioredoxin; PKC-theta-interacting protein; PKCq-interacting protein; testicular tissue protein Li 75; thioredoxin-like 2; Thioredoxin-like protein 2
Gene Aliases: GLRX3; GLRX4; GRX3; GRX4; HUSSY-22; PICOT; TXNL2; TXNL3
UniProt ID: (Human) O76003, (Mouse) Q9CQM9, (Rat) Q9JLZ1
Entrez Gene ID: (Human) 10539, (Mouse) 30926, (Rat) 58815
Molecular Function: oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.