Novus Biologicals
Manufacturer Code:NBP154578H
Catalog # NBP154578H
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | JY2 |
Immunogen | Human recombinant TXNIP |
Conjugate | HRP |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse MonoclonalAntibody
For Research Use Only
Protein Aliases: HHCPA78 THIF thioredoxin binding protein 2 thioredoxin interacting protein Thioredoxin-binding protein 2 thioredoxin-interacting protein upregulated by 125-dihydroxyvitamin D-3 VDUP1EST01027 Vitamin D3 up-regulated protein 1; thioredoxin binding protein 2; Thioredoxin-binding protein 2; Thioredoxin-interacting protein; upregulated by 1,25-dihydroxyvitamin D-3; Vitamin D3 up-regulated protein 1
Gene Aliases: ARRDC6; EST01027; HHCPA78; THIF; TXNIP; VDUP1
UniProt ID: (Human) Q9H3M7
Entrez Gene ID: (Human) 10628
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.