Invitrogen
Manufacturer Code:PA597236
Catalog # PIPA597236
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 1-110 of human TXNDC9. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP binding protein associated with cell differentiation; ATP-binding protein associated with cell differentiation; Protein 1-4; Thioredoxin domain-containing protein 9
Gene Aliases: AI098020; APACD; PHLP3; TXNDC9
UniProt ID: (Human) O14530, (Mouse) Q9CQ79
Entrez Gene ID: (Human) 10190, (Mouse) 98258
Molecular Function:
G-protein modulator
chaperone
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.