Invitrogen
Manufacturer Code:PA546169
Catalog # PIPA546169
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human TXNDC9 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP binding protein; ATP binding protein associated with cell differentiation; ATP-binding protein associated with cell differentiation; ES cell-related protein; Protein 1-4; Thioredoxin domain-containing protein 9
Gene Aliases: AI098020; APACD; fb15e07; PHLP3; TXNDC9; wu:fb15e07; zgc:55522
UniProt ID: (Bovine) O18883, (Human) O14530, (Mouse) Q9CQ79
Entrez Gene ID: (Bovine) 539143, (Rabbit) 100348973, (Dog) 474557, (Horse) 100050502, (Human) 10190, (Guinea pig) 100723650, (Rat) 280671, (Mouse) 98258, (Zebrafish) 336625
Molecular Function:
G-protein modulator
chaperone
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.