Novus Biologicals
Manufacturer Code:NBP189052
Catalog # NBP189052
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:TQDYVVGFTDLGNTDDFTTETLEWRLGSSDILNYSGNLMEPPFQNQKKFGTNFTKLEKKTIRGKKYDSDSDDD |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: APACDProtein 1-4 ATP binding protein associated with cell differentiation ATP-binding protein associated with cell differentiation PHLP3 thioredoxin domain containing 9 thioredoxin domain-containing protein 9; ATP binding protein associated with cell differentiation; ATP-binding protein associated with cell differentiation; Protein 1-4; Thioredoxin domain-containing protein 9
Gene Aliases: APACD; PHLP3; TXNDC9
UniProt ID: (Human) O14530
Entrez Gene ID: (Human) 10190
Molecular Function:
G-protein modulator
chaperone
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.