Invitrogen
Manufacturer Code:PA518297
Catalog # PIPA518297
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (SLHRFVLSQAKDEL) corresponding to the C-terminus amino acids of TXNDC5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: endoplasmic reticulum protein ERp46; Endoplasmic reticulum resident protein 46; endothelial protein disulphide isomerase; ER protein 46; protein disulfide isomerase family A, member 15; thioredoxin domain containing 5 (endoplasmic reticulum); Thioredoxin domain-containing protein 5; thioredoxin related protein; Thioredoxin-like protein p46; TXNDC5 EndoPDI thioredoxin domain containing 5 ERP46 UNQ364 MGC3178 FLJ21353 FLJ90810 thioredoxin related protein endothelial protein disulphide thioredoxin domain containing 5 (endoplasmic reticulum) isomerase Hcc-2 PDIA15 endothelial protein disulphide isomerase protein disulfide isomerase family A member 15 thioredoxin domain containing 5
Gene Aliases: ENDOPDI; ERP46; HCC-2; HCC2; PDIA15; STRF8; TLP46; TXNDC5; UNQ364; UNQ364/PRO700
UniProt ID: (Human) Q8NBS9
Entrez Gene ID: (Human) 81567
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.