Novus Biologicals
Manufacturer Code:NBP2837290.1ML
Catalog # NB031658
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of Human TXNDC2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp434H0311 MGC35026 spermatid-specific thioredoxin-1 SPTRX SPTRX1 thioredoxin domain containing 2 (spermatozoa) thioredoxin domain-containing 2 (spermatozoa) thioredoxin domain-containing protein 2; sperm-specific thioredoxin 1; Spermatid-specific thioredoxin-1; Sptrx-1; testicular tissue protein Li 215; thioredoxin domain containing 2 (spermatozoa); thioredoxin domain-containing 2 (spermatozoa); Thioredoxin domain-containing protein 2
Gene Aliases: SPTRX; SPTRX1; TXNDC2
UniProt ID: (Human) Q86VQ3
Entrez Gene ID: (Human) 84203
Molecular Function: oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.