Novus Biologicals
Manufacturer Code:NBP169614
Catalog # NBP169614
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to TXNDC15(thioredoxin domain containing 15) The peptide sequence was selected from the C terminal of TXNDC15. Peptide sequence STRFGTVAVPNILLFQGAKPMARFNHTDRTLETLKIFIFNQTGIEAKKNV. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2310047H23Rik; C5orf14 chromosome 5 open reading frame 14 disulfide isomerase FLJ22625 thioredoxin domain containing 152310047H23Rik thioredoxin domain-containing protein 15 UNQ335; disulfide isomerase; Thioredoxin domain-containing protein 15
Gene Aliases: C5orf14; TXNDC15; UNQ335; UNQ335/PRO534
UniProt ID: (Human) Q96J42
Entrez Gene ID: (Human) 79770
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.