Novus Biologicals
Manufacturer Code:NBP29345720UL
Catalog # NB028791
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 93-172 of human TXNDC12 (NP_056997.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AG1 AGR1 anterior gradient homolog 1 EC 1.8.4.2 endoplasmic reticulum protein ERp19 Endoplasmic reticulum resident protein 18 Endoplasmic reticulum resident protein 19 endoplasmic reticulum thioredoxin superfamily member 18 kDa ER protein 18 ER protein 19 ERP16 ERp18 ERP18AG1 ERP19 hAG-1 hTLP19 PDIA16 protein disulfide isomerase family A member 16 thioredoxin domain containing 12 (endoplasmic reticulum) thioredoxin domain-containing protein 12 Thioredoxin-like protein p19 TLP19 TLP19hTLP19 TXNDC12 thioredoxin domain containing 12 (endoplasmic reticulum); anterior gradient homolog 1; endoplasmic reticulum protein ERp19; Endoplasmic reticulum resident protein 18; Endoplasmic reticulum resident protein 19; endoplasmic reticulum thioredoxin superfamily member, 18 kDa; ER protein 18; ER protein 19; hTLP19; protein disulfide isomerase family A, member 16; thioredoxin domain containing 12 (endoplasmic reticulum); Thioredoxin domain-containing protein 12; Thioredoxin-like protein p19
Gene Aliases: AG1; AGR1; ERP16; ERP18; ERP19; hAG-1; hTLP19; PDIA16; TLP19; TXNDC12; UNQ713/PRO1376
UniProt ID: (Human) O95881
Entrez Gene ID: (Human) 51060
Molecular Function: oxidoreductase reductase surfactant
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.