Invitrogen
Manufacturer Code:PA599845
Catalog # PIPA599845
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human TXK corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A130089B16Rik BTKL Protein-tyrosine kinase 4 PSCTK5 PTK4 PTK4 protein tyrosine kinase 4 PTK-RL-18 resting lymphocyte kinase RLK TKL Txk TXK tyrosine kinase tyrosine kinase tyrosine-protein kinase TXK; Protein-tyrosine kinase 4; PTK-RL-18; PTK4 protein tyrosine kinase 4; Resting lymphocyte kinase; tyrosine kinase; Tyrosine-protein kinase TXK
Gene Aliases: A130089B16Rik; BTKL; PSCTK5; PTK-RL-18; PTK4; RLK; TKL; TXK
UniProt ID: (Human) P42681, (Mouse) P42682
Entrez Gene ID: (Human) 7294, (Mouse) 22165
Molecular Function: kinase non-receptor tyrosine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.