Invitrogen
Manufacturer Code:PA575953
Catalog # PIPA575953
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 321-366 of Human TXK. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Protein-tyrosine kinase 4; PTK-RL-18; PTK4 protein tyrosine kinase 4; Resting lymphocyte kinase; tyrosine kinase; Tyrosine-protein kinase TXK; Tyrosine-protein kinase TXK Protein-tyrosine kinase 4 Resting lymphocyte kinase TXK PTK4 RLK
Gene Aliases: A130089B16Rik; BTKL; PSCTK5; PTK-RL-18; PTK4; RLK; TKL; TXK
UniProt ID: (Human) P42681, (Mouse) P42682
Entrez Gene ID: (Human) 7294, (Rat) 305311, (Mouse) 22165
Molecular Function: kinase non-receptor tyrosine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.