Abnova
Manufacturer Code:PAB28271
Catalog # 89-129-776
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of recombinant TXK. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Protein Aliases: Protein-tyrosine kinase 4; PTK4 protein tyrosine kinase 4; Resting lymphocyte kinase; tyrosine kinase; Tyrosine-protein kinase TXK
Gene Aliases: BTKL; PSCTK5; PTK4; RLK; TKL; TXK
UniProt ID: (Human) P42681
Entrez Gene ID: (Human) 7294
Molecular Function: kinase non-receptor tyrosine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.