Proteintech
Manufacturer Code:189811AP150UL
Catalog # 50-173-2302
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BTKL Protein tyrosine kinase 4 PSCTK5 PTK4 RLK TKL TXK TXK tyrosine kinase Tyrosine protein kinase TXK; Protein-tyrosine kinase 4; PTK4 protein tyrosine kinase 4; Resting lymphocyte kinase; tyrosine kinase; Tyrosine-protein kinase TXK
Gene Aliases: BTKL; PSCTK5; PTK4; RLK; TKL; TXK
UniProt ID: (Human) P42681
Entrez Gene ID: (Human) 7294
Molecular Function:
kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.