Novus Biologicals
Manufacturer Code:NB10061631
Catalog # NB10061631
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 288 and 338 of human DNA-directed RNA polymerase I subunit RPA43 (TWIST neighbor) using the numbering given in entry NP_001002926.1 (GeneID 221830). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
For Research Use Only
Protein Aliases: DNA-directed RNA polymerase I subunit F; DNA-directed RNA polymerase I subunit RPA43; DNA-directed RNA polymerase I subunit RPA43 TWIST neighbor Twist neighbor protein; Twist neighbor protein
Gene Aliases: POLR1F; TWISTNB
UniProt ID: (Human) Q3B726
Entrez Gene ID: (Human) 221830
Molecular Function: DNA-directed RNA polymerase RNA binding protein nucleic acid binding nucleotidyltransferase transcription factor transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.