Invitrogen
Manufacturer Code:PA599393
Catalog # PIPA599393
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human Twist2 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AMS BBRSAY bHLHa39 Class A basic helix-loop-helix protein 39 dermis expressed 1 dermis-expressed protein 1 Dermo1 Dermo-1 dermo-1 protein FFDD3 SETLSS twist basic helix-loop-helix transcription factor 2 twist family bHLH transcription factor 2 twist homolog 2 TWIST2 twist-related bHLH protein Dermo1 Twist-related protein 2; bHLHa39; Class A basic helix-loop-helix protein 39; dermis expressed 1; Dermis-expressed protein 1; Dermo-1; dermo-1 protein; twist basic helix-loop-helix transcription factor 2; twist homolog 2; twist-related bHLH protein Dermo1; Twist-related protein 2
Gene Aliases: AMS; BBRSAY; BHLHA39; DERMO1; FFDD3; SETLSS; TWIST2
UniProt ID: (Human) Q8WVJ9, (Mouse) Q9D030, (Rat) P97831
Entrez Gene ID: (Human) 117581, (Mouse) 13345, (Rat) 59327
Molecular Function: basic helix-loop-helix transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.