Invitrogen
Manufacturer Code:PA586070
Catalog # PIPA586070
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 1-50 of Human TWIST1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B-HLH DNA binding protein; bHLHa38; charlie chaplin; Class A basic helix-loop-helix protein 38; H-twist; M-twist; Pluridigite; polydactyly EMS; Ska<sup>m10Jus</sup>; twist basic helix-loop-helix transcription factor 1; twist gene homolog 1; twist homolog 1; TWIST homolog of drosophila; Twist-related protein 1; Twist-related protein 1 Class A basic helix-loop-helix protein 38 bHLHa38 H-twist BHLHA38 TWIST
Gene Aliases: AA960487; ACS3; BHLHA38; BPES2; BPES3; CRS; CRS1; CSO; M-Twist; Pde; pdt; SCS; Ska10; Ska<m10Jus>; TWIST; TWIST1
UniProt ID: (Human) Q15672, (Mouse) P26687
Entrez Gene ID: (Human) 7291, (Mouse) 22160
Molecular Function: basic helix-loop-helix transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.