Invitrogen
Manufacturer Code:PA568957
Catalog # PIPA568957
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human TWIST1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B-HLH DNA binding protein; bHLHa38; charlie chaplin; Class A basic helix-loop-helix protein 38; H-twist; M-twist; Pluridigite; polydactyly EMS; Ska<sup>m10Jus</sup>; twist basic helix-loop-helix transcription factor 1; twist gene homolog 1; twist homolog 1; Twist homolog 1 (Drosophila) SCS ACS3 CRS1 BPES2 BPES3 TWIST bHLHa38; TWIST homolog of drosophila; Twist-related protein 1
Gene Aliases: AA960487; ACS3; BHLHA38; BPES2; BPES3; CRS; CRS1; CSO; M-Twist; Pde; pdt; SCS; Ska10; Ska<m10Jus>; TWIST; TWIST1
UniProt ID: (Human) Q15672, (Mouse) P26687
Entrez Gene ID: (Horse) 100272214, (Bovine) 782170, (Human) 7291, (Mouse) 22160, (Rat) 85489
Molecular Function:
basic helix-loop-helix transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.