Invitrogen
Manufacturer Code:PA546889
Catalog # PIPA546889
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived recombit human Twist-1 Met1-His202 |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: B-HLH DNA binding protein; bHLHa38; Class A basic helix-loop-helix protein 38; H-twist; twist basic helix-loop-helix transcription factor 1; twist homolog 1; twist homolog 1p ACS3 BPES2 BPES3 SCS TWIST; TWIST homolog of drosophila; Twist-related protein 1
Gene Aliases: ACS3; BHLHA38; BPES2; BPES3; CRS; CRS1; CSO; SCS; TWIST; TWIST1
UniProt ID: (Human) Q15672
Entrez Gene ID: (Human) 7291
Molecular Function: basic helix-loop-helix transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.