Invitrogen
Manufacturer Code:PA5116093
Catalog # PIPA5116093
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human Twist1 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AA960487 ACS3 B-HLH DNA binding protein bHLHa38 BPES2 BPES3 charlie chaplin class A basic helix-loop-helix protein 38 CRS CRS1 CSO H-twist M-twist p ACS3 Pde pdt Pluridigite polydactyly EMS SCS Ska<m10Jus> Ska<sup>m10Jus</sup> Ska10 TWIST twist basic helix-loop-helix transcription factor 1 twist family bHLH transcription factor 1 twist gene homolog 1 twist homolog 1 TWIST homolog of drosophila Twist1 twist-related protein 1; B-HLH DNA binding protein; bHLHa38; charlie chaplin; Class A basic helix-loop-helix protein 38; H-twist; M-twist; Pluridigite; polydactyly EMS; Ska<sup>m10Jus</sup>; twist basic helix-loop-helix transcription factor 1; twist gene homolog 1; twist homolog 1; TWIST homolog of drosophila; Twist-related protein 1
Gene Aliases: AA960487; ACS3; BHLHA38; BPES2; BPES3; CRS; CRS1; CSO; M-Twist; Pde; pdt; SCS; Ska10; Ska<m10Jus>; TWIST; TWIST1
UniProt ID: (Human) Q15672, (Mouse) P26687
Entrez Gene ID: (Human) 7291, (Mouse) 22160, (Rat) 85489
Molecular Function:
basic helix-loop-helix transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.