Invitrogen
Manufacturer Code:PA5116271
Catalog # PIPA5116271
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 78-104 amino acids from the Central region of human TUSC5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BEC-1 Brain endothelial cell-derived protein 1 C130069F04Rik Dispanin subfamily B member 1 DSPB1 IFITMD3 interferon induced transmembrane protein domain containing 3 interferon-induced transmembrane domain-containing protein D3 located at seventeen p thirteen point three 1 LOST1 Protein located at seventeen-p-thirteen point three 1 Trafficking regulator of GLUT4 1 Trarg1 Tumor suppressor candidate 5 tumor suppressor candidate 5 homolog Tusc5; Dispanin subfamily B member 1; DSPB1; interferon induced transmembrane protein domain containing 3; Interferon-induced transmembrane domain-containing protein D3; located at seventeen p thirteen point three 1; Protein located at seventeen-p-thirteen point three 1; Trafficking regulator of GLUT4 1; Tumor suppressor candidate 5
Gene Aliases: DSPB1; IFITMD3; LOST1; TRARG1; TUSC5
UniProt ID: (Human) Q8IXB3
Entrez Gene ID: (Human) 286753
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.