Abnova
Manufacturer Code:MAB1790
Catalog # 89-110-629
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 10 |
Immunogen | Recombinant protein corresponding to amino acids 1-200 of Yeast TUP1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against recombinant TUP1.
For Research Use Only
Protein Aliases: AAR1; AER2; AMM1; chromatin-silencing transcriptional regulator TUP1; CRT4; CYC9; FLK1; Flocculation suppressor protein; General transcriptional corepressor TUP1; Glucose repression regulatory protein TUP1; Repressor AER2; ROX4; SFL2; UMR7
Gene Aliases: AAR1; AER2; AMM1; CRT4; CYC9; FLK1; ROX4; SFL2; TUP1; UMR7; YCR084C; YCR84C
UniProt ID: (Baker's yeast) P16649
Entrez Gene ID: (Baker's yeast) 850445
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.