Invitrogen
Manufacturer Code:MA112683
Catalog # 01-674-035
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Baker's yeast |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 10 |
Immunogen | Recombit protein corresponding to the residues 1-200 of yeast TUP1 protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: chromatin-silencing transcriptional regulator TUP1; Flocculation suppressor protein; General transcriptional corepressor TUP1; Glucose repression regulatory protein TUP1; Repressor AER2; YCR084C AAR1 AER2 AMM1 CRT4 CYC9 FLK1 ROX4 SFL2 UMR7
Gene Aliases: AAR1; AER2; AMM1; CRT4; CYC9; FLK1; ROX4; SFL2; TUP1; UMR7; YCR084C; YCR84C
UniProt ID: (Baker's yeast) P16649
Entrez Gene ID: (Baker's yeast) 850445
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.