OriGene
Manufacturer Code:TA504148
Catalog # 50-167-4895
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1A10 |
Immunogen | Full length human recombit protein of human TULP3 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: tubby like protein 3; Tubby-like protein 3; Tubby-related protein 3; TUBL3
Gene Aliases: 2310022L06Rik; AI316887; TUBL3; TULP3
UniProt ID: (Human) O75386, (Mouse) O88413
Entrez Gene ID: (Dog) 611832, (Human) 7289, (Mouse) 22158
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.