Invitrogen
Manufacturer Code:PA541449
Catalog # PIPA541449
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human TULP2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cancer testis antigen 65; Cancer/testis antigen 65; CT65; Protein P4-6; tubby like protein 2; Tubby like protein 2 TUBL2 CT65; Tubby-like protein 2; Tubby-related protein 2
Gene Aliases: CT65; Pdet; TUBL2; TULP2
UniProt ID: (Human) O00295, (Mouse) P46686
Entrez Gene ID: (Horse) 100051453, (Human) 7288, (Bovine) 505350, (Dog) 484392, (Guinea pig) 100727285, (Mouse) 56734, (Rat) 361576
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.