Novus Biologicals
Manufacturer Code:NBP215151
Catalog # NBP215151
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the C-terminus region of human TULP2. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cancer testis antigen 65; Cancer/testis antigen 65; Cancer/testis antigen 65 CT65cancer testis antigen 65 tubby like protein 2 Tubby-like protein 2 TUBL2tubby-related protein 2; CT65; Tubby-like protein 2; Tubby-related protein 2
Gene Aliases: CT65; TUBL2; TULP2
UniProt ID: (Human) O00295
Entrez Gene ID: (Human) 7288
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.