Proteintech
Manufacturer Code:189711AP150UL
Catalog # 50-173-2295
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TULP1 Fusion Protein Ag5598 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: RP14 tubby like protein 1 Tubby related protein 1 TUBL1 TULP1; tubby like protein 1 like; Tubby-like protein 1; Tubby-related protein 1
Gene Aliases: LCA15; RP14; TUBL1; TULP1; Tulp1l
UniProt ID: (Human) O00294, (Mouse) Q9Z273
Entrez Gene ID: (Human) 7287, (Mouse) 22157, (Rat) 309900
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.