Novus Biologicals
Manufacturer Code:NBP236751
Catalog # NBP236751
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | CL2242 |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:AVDTYIPVPARDLEKPFLLPVEAVYSVPGRGTVVTGTLERGILKKGDECELLGHSKNIRTVVTGIEMFHKSLERAEAGDNLGALVRGLKREDLRRGLVMVKPGSI |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: EF-Tu; EFTu EF-Tu EFTUCOXPD4 EF-TuMT elongation factor Tu mitochondrial P43 Tu translation elongation factor mitochondrial; Elongation factor Tu, mitochondrial; P43
Gene Aliases: COXPD4; EF-TuMT; EFTU; P43; TUFM
UniProt ID: (Human) P49411
Entrez Gene ID: (Human) 7284
Molecular Function: G-protein RNA binding protein enzyme modulator hydrolase nucleic acid binding translation elongation factor translation factor translation initiation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.