Invitrogen
Manufacturer Code:PA544906
Catalog # PIPA544906
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human TUFM |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EF-Tu; Elongation factor Tu, mitochondrial; P43; Tu translation elongation factor mitochondrial COXPD4 EF-TuMT EFTU P43; tufm {ECO:0000250|UniProtKB:P49411}
Gene Aliases: 2300002G02Rik; C76308; C76389; COXPD4; D250; EF-Tu; EF-TuMT; EFTU; fi06f04; P43; TUFM; wu:fi06f04; zgc:110766
UniProt ID: (Bovine) P49410, (Human) P49411, (Mouse) Q8BFR5, (Rat) P85834
Entrez Gene ID: (Bovine) 281556, (Dog) 479796, (Human) 7284, (Rabbit) 100344982, (Horse) 100064341, (Mouse) 233870, (Guinea pig) 100727064, (Rat) 293481, (Zebrafish) 541378
Molecular Function: G-protein RNA binding protein enzyme modulator hydrolase nucleic acid binding translation elongation factor translation factor translation initiation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.