Invitrogen
Manufacturer Code:PA544635
Catalog # PIPA544635
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human TUBE1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Epsilon-tubulin; epsilon-tubulin 1; Tubulin epsilon 1 FLJ22589 TUBE dJ142L7.2; Tubulin epsilon chain; tubulin, epsilon 1
Gene Aliases: 2310061K05Rik; AI551343; dJ142L7.2; TUBE; TUBE1
UniProt ID: (Human) Q9UJT0, (Mouse) Q9D6T1
Entrez Gene ID: (Human) 51175, (Dog) 612390, (Rabbit) 100340587, (Horse) 100072438, (Mouse) 71924, (Rat) 361856, (Guinea pig) 100727567
Molecular Function: cytoskeletal protein microtubule family cytoskeletal protein tubulin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.