Invitrogen
Manufacturer Code:PA5103476
Catalog # PIPA5103476
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human TUBE1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2310061K05Rik AI551343 dJ142L7.2 Epsilon-tubulin epsilon-tubulin 1 FLJ22589 FLJ44203 RP1-142L7.1 TUBE Tube1 Tubulin tubulin epsilon 1 tubulin epsilon chain tubulin epsilon 1; Epsilon-tubulin; Tubulin epsilon chain; tubulin, epsilon 1
Gene Aliases: 2310061K05Rik; AI551343; dJ142L7.2; TUBE; TUBE1
UniProt ID: (Human) Q9UJT0, (Mouse) Q9D6T1
Entrez Gene ID: (Human) 51175, (Mouse) 71924
Molecular Function:
cytoskeletal protein
microtubule family cytoskeletal protein
tubulin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.