OriGene
Manufacturer Code:TA503907
Catalog # 50-167-4684
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4D8 |
Immunogen | Human recombit protein fragment corresponding to amino acids 150-446 of human TUBAL3 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Tubulin alpha chain-like 3; tubulin alpha like 3; tubulin, alpha-like 3
Gene Aliases: TUBAL3
UniProt ID: (Human) A6NHL2
Entrez Gene ID: (Human) 79861
Molecular Function:
cytoskeletal protein
microtubule family cytoskeletal protein
tubulin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.