Invitrogen
Manufacturer Code:PA540723
Catalog # PIPA540723
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human TUB |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: retinal degeneration 5; tubby bipartite transcription factor; tubby homolog; Tubby homolog (mouse) rd5; tubby homologue; Tubby protein; Tubby protein homolog
Gene Aliases: Rd5; RDOB; TUB
UniProt ID: (Human) P50607, (Mouse) P50586, (Rat) O88808
Entrez Gene ID: (Horse) 100070898, (Dog) 485377, (Human) 7275, (Rabbit) 100342264, (Guinea pig) 100732995, (Mouse) 22141, (Rat) 25609
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.