Proteintech
Manufacturer Code:269731AP150UL
Catalog # 50-173-5521
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TTYH1 Fusion Protein Ag25290 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hTTY1; hTTY1 Protein tweety homolog 1 TTYH1 tweety homolog 1 (Drosophila); mTTY1; Protein tweety homolog 1; tweety homolog 1
Gene Aliases: 4930459B04Rik; 6330408P11Rik; mTTY1; tty; TTY1; TTYH1
UniProt ID: (Human) Q9H313, (Mouse) Q9D3A9, (Rat) P0C5X8
Entrez Gene ID: (Human) 57348, (Mouse) 57776, (Rat) 292597
Molecular Function: anion channel ion channel transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.