Abnova
Manufacturer Code:H00051567K
Catalog # 89-200-637
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human TTRAP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human TTRAP peptide using ARM Technology.
For Research Use Only
Protein Aliases: 5'-Tyr-DNA phosphodiesterase; 5'-tyrosyl-DNA phosphodiesterase; AD022 EAP2 MGC111021 MGC9099 dJ30M3.3; EAPII; ETS1-associated protein 2; ETS1-associated protein II; TRAF and TNF receptor associated protein; TRAF and TNF receptor-associated protein; Tyr-DNA phosphodiesterase 2; Tyrosyl-DNA phosphodiesterase 2; Tyrosyl-RNA phosphodiesterase; VPg unlinkase
Gene Aliases: AD-022; AD022; dJ30M3.3; EAP2; EAPII; hTDP2; TDP2; TTRAP
UniProt ID: (Human) O95551
Entrez Gene ID: (Human) 51567
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.