Abnova
Manufacturer Code:H00007274M01A
Catalog # 89-105-681
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 7B5 |
Immunogen | TTPA (NP_000361.1179 a.a. &bsim 278 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant TTPA.
For Research Use Only
Protein Aliases: Alpha-tocopherol transfer protein; Alpha-TTP; alphaTTP; ATTP; AVED; tocopherol (alpha) transfer protein (ataxia (Friedreich-like) with vitamin E deficiency); TTP1
Gene Aliases: alphaTTP; ATTP; AVED; TPP1; TTP1; TTPA
UniProt ID: (Human) P49638
Entrez Gene ID: (Human) 7274
Molecular Function: transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.