Proteintech
Manufacturer Code:270811AP150UL
Catalog # 50-173-5555
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TTPA Fusion Protein Ag24177 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha TTP; Alpha TTP alphaTTP ATTP AVED TPP1 TTP1 TTPA; Alpha-tocopherol transfer protein; Alpha-TTP; tocopherol (alpha) transfer protein (ataxia (Friedreich-like) with vitamin E deficiency); Tocopherol transfer protein alpha
Gene Aliases: alpha-TTP; alphaTTP; ATTP; AVED; TPP1; TTP; TTP1; TTPA
UniProt ID: (Human) P49638, (Rat) P41034, (Mouse) Q8BWP5
Entrez Gene ID: (Human) 7274, (Rat) 25571, (Mouse) 50500
Molecular Function:
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.