Invitrogen
Manufacturer Code:PA527285
Catalog # PIPA527285
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 423 of Human TTLL1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bM121M7.1 (novel protein (ortholog of human tubulin tyrosine ligase-like 1 (TTLL1)), isoform 1); C22orf7 HS323M22B PGs3 catalytic subunit of neural tubulin polyglutamylase probable tubulin polyglutamylase TTLL1 tubulin polyglutamylase complex subunit 3 tubulin--tyrosine ligase-like protein 1 tubulin-tyrosine ligase; catalytic subunit of neural tubulin polyglutamylase; p49; PGs3; Probable tubulin polyglutamylase TTLL1; Tubulin polyglutamylase complex subunit 3; tubulin tyrosine ligase-like family member 1; tubulin tyrosine ligase-like family, member 1; Tubulin--tyrosine ligase-like protein 1; tubulin-tyrosine ligase
Gene Aliases: 6330444E16Rik; AV014541; C22orf7; HS323M22B; TTLL1
UniProt ID: (Human) O95922, (Mouse) Q91V51
Entrez Gene ID: (Human) 25809, (Mouse) 319953
Molecular Function:
cytoskeletal protein
ligase
microtubule family cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.