Abnova
Manufacturer Code:H00007272M02
Catalog # 89-001-034
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4G9 |
Immunogen | TTK (AAH00633 1 a.a. ~ 176 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant TTK.
For Research Use Only
Protein Aliases: cancer/testis antigen 96; Dual specificity protein kinase TTK; ESK; FLJ38280; monopolar spindle 1 kinase; monopolar spindle 1-like 1; MPS1; MPS1L1; phosphotyrosine picked threonine kinase; Phosphotyrosine picked threonine-protein kinase; PYT
Gene Aliases: CT96; ESK; MPH1; MPS1; MPS1L1; PYT; TTK
UniProt ID: (Human) P33981
Entrez Gene ID: (Human) 7272
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.