Novus Biologicals
Manufacturer Code:NB100462
Catalog # NB100462
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residues 700 and 750 of human TTK using the numbering given Swiss-Prot in entry P33981 (GeneID 7272). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cancer/testis antigen 96; CT96 dual specificity protein kinase TTK EC 2.7.12.1 ESK FLJ38280 monopolar spindle 1-like 1 MPS1 MPS1L1PYTcancer/testis antigen 96 phosphotyrosine picked threonine kinase Phosphotyrosine picked threonine-protein kinase TTK protein kinase; Dual specificity protein kinase TTK; monopolar spindle 1 kinase; monopolar spindle 1-like 1; phosphotyrosine picked threonine kinase; Phosphotyrosine picked threonine-protein kinase; PYT
Gene Aliases: CT96; ESK; MPH1; MPS1; MPS1L1; PYT; TTK
UniProt ID: (Human) P33981
Entrez Gene ID: (Human) 7272
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.