Invitrogen
Manufacturer Code:PA568792
Catalog # PIPA568792
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the n terminal region of mouse TTF1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BCH BHC NK-2 NKX2.1 NKX2A TEBP TITF1 TTF-1 TTF1 NK-2 homolog A homeobox protein NK-2 homolog A homeobox protein Nkx-2.1 thyroid nuclear factor 1 thyroid transcription factor 1; RNA polymerase I termination factor; Transcription termination factor 1; Transcription termination factor I; transcription termination factor, RNA polymerase I; TTF-1; TTF-I
Gene Aliases: AV245725; RGD1565673; TTF-1; TTF-I; Ttf1
UniProt ID: (Mouse) Q62187
Entrez Gene ID: (Rat) 499766, (Mouse) 22130
Molecular Function: DNA binding protein nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.