Invitrogen
Manufacturer Code:PA543307
Catalog # PIPA543307
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human TTC8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Bardet-Biedl syndrome 8 protein; Bardet-Biedl syndrome 8 protein homolog; Bardet-Biedl syndrome type 8; Tetratricopeptide repeat domain 8 BBS8 RP51; Tetratricopeptide repeat protein 8; TPR repeat protein 8
Gene Aliases: 0610012F22Rik; AV001447; BBS8; RP51; TTC8
UniProt ID: (Human) Q8TAM2, (Mouse) Q8VD72
Entrez Gene ID: (Rabbit) 100350688, (Human) 123016, (Dog) 480413, (Horse) 100052922, (Rat) 299246, (Guinea pig) 100723278, (Mouse) 76260
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.