Novus Biologicals
Manufacturer Code:NBP15510820UL
Catalog # NBP15510820
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to TTC8(tetratricopeptide repeat domain 8) The peptide sequence was selected from the N terminal of TTC8. Peptide sequence ENAIAQVPRPGTSLKLPGTNQTGGPSQAVRPITQAGRPITGFLRPSTQSG. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Bardet-Biedl syndrome 8 protein; Bardet-Biedl syndrome 8 protein BBS8Bardet-Biedl syndrome type 8 RP51 tetratricopeptide repeat domain 8 tetratricopeptide repeat protein 8 TPR repeat protein 8; Bardet-Biedl syndrome type 8; Tetratricopeptide repeat protein 8; TPR repeat protein 8
Gene Aliases: BBS8; RP51; TTC8
UniProt ID: (Human) Q8TAM2
Entrez Gene ID: (Human) 123016
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.